missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EI24 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81820AF405
This item is not returnable.
View return policy
Description
EI24 Polyclonal antibody specifically detects EI24 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| EI24 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 405 | |
| EPG4, etoposide induced 2.4 mRNA, p53-induced gene 8 protein, PIG8etoposide-induced protein 2.4 homolog, TP53I8, tumor protein p53 inducible protein 8 | |
| EI24 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human EI24. The immunogen is located within the last 50 amino acids of EI24. Amino Acid Squence: STSAEKFPSPHPSPAKLKA | |
| 0.1 mL | |
| Apoptosis | |
| 9538 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction