Learn More
EI24 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20456-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.Specifications
| EI24 | |
| Polyclonal | |
| Unconjugated | |
| EI24 | |
| EI24, etoposide induced 2.4 mRNA, p53 induced gene 8 protein, p53-induced factor Ei24, PIG8, TP53I8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13663, 300514, 9538 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14681, Q4KM77, Q61070 | |
| EI24 | |
| EI24 Fusion Protein Ag14169 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |