missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eicosanoic-2,2-d2 Acid, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Eicosanoic Acid-2,2-D2 |
| CAS | 232600-70-3 |
| Molecular Formula | C20 D2 H38 O2 |
| Quantity | 1 g |
| Synonym | Eicosanoic-2,2-d2 acid (9 CI), Eicosanoic Acid-2,2-D2, Eicosanoic acid-d2, Arachidic-2,2-d2 acid, Arachidic acid-2,2-d2, Arachidic acid-α-d2 |
| InChI Formula | InChI=1 S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)/i19D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCCCCCCCCCCCC)C(=O)O |
| IUPAC Name | 2,2-dideuterioicosanoic acid |
| Molecular Weight (g/mol) | 314.5427 |
| Formula Weight | 314.3154 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction