missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EID3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531556
This item is not returnable.
View return policy
Description
Recommended positive controls: IMR32. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Tissue-specific component of the SMC5-SMC6 complex, a complex involved in repair of DNA double-strand breaks by homologous recombination. The complex may promote sister chromatid homologous recombination by recruiting the SMC1-SMC3 cohesin complex to double-strand breaks. The complex is required for telomere maintenance via recombination and mediates sumoylation of shelterin complex (telosome) components. [UniProt].
Specifications
| EID3 | |
| Polyclonal | |
| Unconjugated | |
| EID3 | |
| 1700027M21Rik; E1A-like inhibitor of differentiation 3; EID-1-like inhibitor of differentiation 3; Eid3; EID-3; eid3 {ECO:0000312; EMBL:AAH97404.1}; EP300 interacting inhibitor of differentiation 3; EP300-interacting inhibitor of differentiation 3; non-SMC element 4 homolog B; non-structural maintenance of chromosomes element 4 homolog B; NS4EB; NSE4B; NSMCE4B; testis tissue sperm-binding protein Li 96mP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 493861 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N140 | |
| EID3 | |
| Recombinant protein encompassing a sequence within the center region of human EID3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction