missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF1AX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117033
This item is not returnable.
View return policy
Description
Positive Samples: SKOV3, HT-29, MCF7, NCI-H460, Mouse brain, Mouse kidney Immunogen sequence: KNRRRGKNEN ESEKRELVFK EDGQEYAQVI KMLGNGRLEA MCFDGVKRLC HIRGKLRKKV WINTSDIILV GLRDYQDNKA DVILKYNADE ARSLKAYGEL PEHAKINETD TFGPGDDDEI QFDDIGDDDE DIDDI.
This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
Specifications
| eIF1AX | |
| Polyclonal | |
| Unconjugated | |
| EIF1AX | |
| 1500010B24Rik; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; EIF1AP1; EIF1AX; Eif1ay; EIF4C; eIF-4C; eukaryotic translation initiation factor 1A X-linked; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1964, 302697, 66235 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P47813, Q8BMJ3 | |
| EIF1AX | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-144 of human EIF1AX (NP_0014031). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction