missing translation for 'onlineSavingsMsg'
Learn More

EIF1AY Antibody (OTI2G2), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-71417UV

Product Code. 18125725

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EIF1A Monoclonal specifically detects EIF1A in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EIF1A
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
EIF1AX
Recombinant protein fragment corresponding to amino acids 2-144 of human EIF1AY (NP_004672) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot
OTI2G2
Western Blot
eIF-1A, EIF1AP1, eIF-4C, EIF4C, eukaryotic translation initiation factor 1A, X chromosome, eukaryotic translation initiation factor 1A, X-chromosomal, eukaryotic translation initiation factor 1A, Y chromosome, eukaryotic translation initiation factor 1A, Y-chromosomal, Eukaryotic translation initiation factor 4C, FLJ45762, Putative eukaryotic translation initiation factor 1A
Mouse
Affinity Purified
DNA replication Transcription Translation and Splicing
1964
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.