missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF1AY Monoclonal Antibody (OTI2G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526885
This item is not returnable.
View return policy
Description
eIF1AY Monoclonal Antibody for Western Blot
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
Specifications
| eIF1AY | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14602 | |
| EIF1AY | |
| Human recombinant protein fragment corresponding to amino acids 2-144 of EIF1AY produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI2G2 | |
| Unconjugated | |
| EIF1AY | |
| 1500010B24Rik; Ab1-287; AI426898; EIF1A; eIF-1A; eIF-1A X isoform; eIF-1A Y isoform; EIF1AP1; EIF1AX; EIF1AY; EIF4C; eIF-4C; Eukaryotic translation initiation factor 1A; eukaryotic translation initiation factor 1A, X chromosome; eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, X-linked; eukaryotic translation initiation factor 1A, Y chromosome; eukaryotic translation initiation factor 1A, Y-chromosomal; eukaryotic translation initiation factor 1A, Y-linked; eukaryotic translation initiation factor 4C; Liver regeneration-related protein LRRG048; Putative eukaryotic translation initiation factor 1A; RGD1560198; X-linked eukaryotic translation initiation factor 1A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9086 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction