Learn More
EIF1AY Mouse anti-Human, Clone: OTI2G2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807511
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5′ end of capped RNA.Specifications
| EIF1AY | |
| Monoclonal | |
| Unconjugated | |
| EIF1AY | |
| eIF-4C | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9086 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2G2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O14602 | |
| EIF1AY | |
| Human recombit protein fragment corresponding to amino acids 2-144 of human EIF1AY produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.