Learn More
eIF2A, p Ser51 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NB100-81896-0.025MG
 View more versions of this product
Product Code. 18211195
  • € 215.00 / 25µg
Description
eIF2A Polyclonal specifically detects eIF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eIF2A (p Ser51)
Polyclonal
Unconjugated
phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD )
Rabbit
38 kDa
0.025 mg
DNA Repair, Phospho Specific, Translation Control, Unfolded Protein Response
83939
Mouse, Rat, Human
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
P05198
EIF2A
The antiserum was produced against synthesized phosphopeptide derived from human eIF2. around the phosphorylation site of serine 51 (E-L-SP-R-R).
Affinity Purified
RUO
Primary
eIF2. (specific to Phospho-Ser51) detects endogenous levels of eIF2. only when phosphorylated at serine 51.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.