missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EIF2A Recombinant Rabbit Monoclonal Antibody (8V8H7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538202
This item is not returnable.
View return policy
Description
Positive Samples: BxPC-3, Mouse liver, Rat lung Immunogen sequence: ALRNKPITNS KLHEEEPPQN MKPQSGNDKP LSKTALKNQR KHEAKKAAKQ EARSDKSPDL APTPAPQSTP RNTVSQSISG DPEIDKKIKN LKKKLKAIEQ L.
EIF2A is a 65-kD protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes.
Specifications
| EIF2A | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BJW6, Q9BY44 | |
| EIF2A | |
| A synthetic peptide corresponding to a sequence within amino acids 450-550 of human eIF2A (Q9BY44). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 8V8H7 | |
| Unconjugated | |
| EIF2A | |
| 0910001O23Rik; 2410026C18Rik; 35kDa; 65 kDa eukaryotic translation initiation factor 2A; CDA02; D030048D22; D3Ertd194e; EIF2; EIF-2; EIF2A; eIF-2A; eIF2alpha; eIF-2alpha; eIF-2-alpha; EIF2S1; EIF2S1 protein; eukaryotic translation initiation factor 2 subunit 1; Eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; eukaryotic translation initiation factor 2A, 65kDa; Eukaryotic translation initiation factor 2A, N-terminally processed; eukaryotic translation initiation factor subunit 1-like protein; initiation factor 2 alpha; MST089; MSTP004; MSTP089 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 229317, 502531, 83939 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction