missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EIF2B4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530292
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Chimpanzee (97%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| EIF2B4 | |
| Polyclonal | |
| Unconjugated | |
| EIF2B4 | |
| DKFZp586J0119; EIF2B; EIF-2B; eIF-2B GDP-GTP exchange factor subunit delta; EIF2B4; Eif2bd; EIF2Bdelta; eukaryotic translation initiation factor 2B subunit 4 delta; eukaryotic translation initiation factor 2B subunit delta; eukaryotic translation initiation factor 2B, subunit 4 delta; eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa; Jgr1a; translation initiation factor eIF-2b delta subunit; translation initiation factor eIF-2B subunit delta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117019, 13667, 8890 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.61 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q61749, Q63186, Q9UI10 | |
| EIF2B4 | |
| Recombinant protein encompassing a sequence within the center region of human EIF2B delta. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction