Learn More
eIF2B3 Antibody (OTI1A4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71397AF750
Product Code. 18112414
  • € 751.00 / 0.10mL
Description
eIF2B3 Monoclonal specifically detects eIF2B3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eIF2B3
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
EIF2B3
Full length recombinant protein of human eIF2B3 (NP_065098) produced in HEK293T cells.
Primary
Human, Mouse, Rat, Canine
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1A4
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
EIF-2B, eIF-2B GDP-GTP exchange factor subunit gamma, EIF2Bgamma, eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD), eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa, translation initiation factor eIF-2B subunit gamma
Mouse
0.1 mL
8891
Store at 4°C in the dark.