missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2B3 Antibody (OTI1A4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71397B
This item is not returnable.
View return policy
Description
eIF2B3 Monoclonal specifically detects eIF2B3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| eIF2B3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| EIF2B3 | |
| Full length recombinant protein of human eIF2B3 (NP_065098) produced in HEK293T cells. | |
| 0.1 mL | |
| 8891.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1A4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EIF-2B, eIF-2B GDP-GTP exchange factor subunit gamma, EIF2Bgamma, eukaryotic translation initiation factor 2B, subunit 3 (gamma, 58kD), eukaryotic translation initiation factor 2B, subunit 3 gamma, 58kDa, translation initiation factor eIF-2B subunit gamma | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction