Learn More
EIF2B3 Monoclonal Antibody (OTI2D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA503568
Encompass_Preferred
 View more versions of this product
Product Code. 17559551
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
EIF2B3 catalyzes the exchange of eukaryotic initiation factor 2-bound GDP for GTP. Mutations in each of the five subunits of translation initiation factor eIF2B, including subunit 3, can cause leukoencephalopathy with vanishing white matter. EIF2B3 has also been identified as a cofactor of hepatitis C virus internal ribosome entry site-mediated translation.
Specifications
Specifications
EIF2B3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NR50
EIF2B3
Full length human recombit protein of human EIF2B3 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2D2
Unconjugated
EIF2B3
EIF-2B, EIF2Bgamma
Mouse
Affinity Chromatography
RUO
8891
-20° C, Avoid Freeze/Thaw Cycles