Learn More
EIF2B3 Mouse anti-Human, Clone: OTI3H3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503681
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
EIF2B3 catalyzes the exchange of eukaryotic initiation factor 2-bound GDP for GTP. Mutations in each of the five subunits of translation initiation factor eIF2B, including subunit 3, can cause leukoencephalopathy with vanishing white matter. EIF2B3 has also been identified as a cofactor of hepatitis C virus internal ribosome entry site-mediated translation.Specifications
| EIF2B3 | |
| Monoclonal | |
| Unconjugated | |
| EIF2B3 | |
| EIF-2B, EIF2Bgamma | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8891 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NR50 | |
| EIF2B3 | |
| Full length human recombit protein of human EIF2B3 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.