missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3D Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97518AF405
This item is not returnable.
View return policy
Description
EIF3D Polyclonal specifically detects EIF3D in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| EIF3D | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 8664 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 405 | |
| eIF3d, eIF3-p66, EIF3S7eIF3 p66, eIF-3-zeta, eIF3-zeta, Eukaryotic translation initiation factor 3 subunit 7, eukaryotic translation initiation factor 3 subunit D, eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa, eukaryotic translation initiation factor 3, subunit D, MGC126526, MGC17258, translation initiation factor eIF3 p66 subunit | |
| Produced in rabbits immunized with E. coli-derived Human EIF3D fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction