missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3D Monoclonal Antibody (OTI14D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808199
This item is not returnable.
View return policy
Description
eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.Specifications
| EIF3D | |
| Monoclonal | |
| Unconjugated | |
| EIF3D | |
| EIF3S7, eIF3-p66, eIF3-zeta | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8664 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI14D6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15371 | |
| EIF3D | |
| Full length human recombit protein of human EIF3D produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction