missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3g Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583585
This item is not returnable.
View return policy
Description
Immunogen sequence: ICKGDHWTTR CPYKDTLGPM QKELAEQLGL STGEKEKLPG ELEPVQATQN KTGKYVPPSL RDGASRRGES MQPN Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 97%.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of posttermination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. This subunit can bind 18S rRNA.
Specifications
| eIF3g | |
| Polyclonal | |
| Unconjugated | |
| EIF3G | |
| 44kDa; D0Jmb4; eiF3 delta; eIF3 p42; eIF3 p42 {ECO:0000255; eIF3 p44; eIF3 p44 {ECO:0000255; eIF-3 RNA-binding subunit; eIF-3 RNA-binding subunit {ECO:0000255; eIF-3-delta; eIF3-delta; eIF-3-delta {ECO:0000255; EIF3G; eIF3g {ECO:0000255; Eif3p42; EIF3-P42; eIF3-p44; EIF3S4; eukaryotic translation initiation factor 3 RNA-binding subunit; eukaryotic translation initiation factor 3 RNA-binding subunit {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 4; eukaryotic translation initiation factor 3 subunit 4 {ECO:0000255; eukaryotic translation initiation factor 3 subunit G; eukaryotic translation initiation factor 3 subunit G {ECO:0000255; eukaryotic translation initiation factor 3, subunit 4 (delta); eukaryotic translation initiation factor 3, subunit 4 (delta, 44 kDa); eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD); eukaryotic translation initiation factor 3 subunit p42; eukaryotic translation initiation factor 3, subunit 4 delta; eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa; eukaryotic translation initiation factor 3, subunit G; HAMAP-Rule:MF_03006}; p44; similar to eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa; translation initiation factor eIF3-p44; TU-189B2; Unknown (protein for MGC:128341); zgc:56553 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8666 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75821 | |
| EIF3G | |
| Recombinant protein corresponding to Human EIF3G. Recombinant protein control fragment (Product #RP-99099). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction