missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3j Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117929
This item is not returnable.
View return policy
Description
eIF3j Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004; Fraser et al., 2007).
Specifications
| eIF3j | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O75822 | |
| EIF3J | |
| E. coli-derived Human EIF3S1/EIF3J fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| EIF3J | |
| 1810016I04Rik; 2700079K05Rik; AA409117; AA409446; C78575; eIF3 p35; eIF3 p35 {ECO:0000255; eIF-3-alpha; eIF3-alpha; eIF-3-alpha {ECO:0000255; eIF-3-alpha-A; eIF-3-alpha-B; Eif3j; eIF3j {ECO:0000255; Eif3j1; Eif3j-1; Eif3j2; Eif3j-2; eIF3j-A; eIF3j-B; eIF3-p35; EIF3S1; Eif3s1-1; Eif3s1-2; ENSMUSG00000043424; Eukaryotic translation initiation factor 3 subunit 1; eukaryotic translation initiation factor 3 subunit 1 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 1-A; Eukaryotic translation initiation factor 3 subunit 1-B; eukaryotic translation initiation factor 3 subunit J; eukaryotic translation initiation factor 3 subunit J {ECO:0000255; Eukaryotic translation initiation factor 3 subunit J-A; Eukaryotic translation initiation factor 3 subunit J-B; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha; eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 3, subunit J; eukaryotic translation initiation factor 3, subunit J1; eukaryotic translation initiation factor 3, subunit J2; Gm9781; HAMAP-Rule:MF_03009}; PRO0391 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8669 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction