Learn More
Invitrogen™ eIF3l Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597314
Description
Immunogen sequence: MSYPADDYES EAAYDPYAYP SDYDMHTGDP KQDLAYERQY EQQTYQVIPE VIKNFIQYFH KTVSDLIDQK VYELQASRVS SDVIDQKVYE IQDIYENSWT KLTERFFKNT PWPEAEAIAP QVGNDAVFLI LYKELYYRHI YAKVSGGPSL EQRFESYYNY CNLFNYILNA DGPAPLELPN QWLWDIIDEF IYQFQSFSQY RCKTAKKSEE EIDFLRSNPK IWNVHSVLNV; Positive Samples: HepG2, HT-1080, NIH/3T3, Mouse heart, Mouse pancreas, Mouse liver; Cellular Location: Cytoplasm.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis (PubMed:17581632, PubMed:25849773, PubMed:27462815). The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation (PubMed:17581632). The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression (PubMed:25849773). [UniProt].
Specifications
| eIF3l | |
| Polyclonal | |
| Unconjugated | |
| EIF3L | |
| 0610011H21Rik; 66 kDa tyrosine-rich heat shock protein; 67 kDa polymerase-associated factor; D15N1e; eIEF associated protein HSPC021; EIF3EIP; Eif3ip; EIF3L; EIF3S11; Eif3s6ip; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit L; eukaryotic translation initiation factor 3, subunit 6 interacting protein; eukaryotic translation initiation factor 3, subunit L; HSP-66Y; HSPC021; HSPC025; MSTP005; Paf67 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 223691, 300069, 51386 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8QZY1, Q9Y262 | |
| EIF3L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human EIF3L (NP_0571751). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.