missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EIF3M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100244
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total EIF3M.
This gene encodes a protein that is part of the eurkaryotic translation initiation factor 3 complete (eIF-3) required for protein synthesis. Elevated levels of the encoded protein are present in cancer cell lines. Inactivation of the encoded protein has been shown to interfere with translation of herpes virus mRNAs by preventing the association of mRNAs with the ribosomes. A pseudogene of this gene is located on the X chromosome.
Specifications
| EIF3M | |
| Polyclonal | |
| Unconjugated | |
| EIF3M | |
| B5; B5 receptor; dendritic cell protein; dendritic cell protein GA17; eIF3m; Eukaryotic translation initiation factor 3 subunit M; eukaryotic translation initiation factor 3, subunit M; fetal lung protein B5; FLJ29030; GA17; HFLB5; hFL-B5; PCI domain containing 1 (herpesvirus entry mediator); PCI domain-containing protein 1; PCID1; PNAS-125; RGD1565840; Tango7; transport and golgi organization 7 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10480, 295975, 98221 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7L2H7, Q99JX4 | |
| EIF3M | |
| A synthesized peptide derived from human EIF3M(Accession Q7L2H7), corresponding to amino acid residues T338-T374. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction