missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517258
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.
Specifications
| eIF4A1 | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P60842, P60843 | |
| EIF4A1 | |
| Synthetic peptide corresponding to residues surrounding Phe52 of human eIF4A. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| EIF4A1 | |
| ATP-dependent RNA helicase eIF4A-1; BM-010; DDX2A; Eif4; EIF4A; EIF-4A; Eif4a1; eIF-4A-I; eIF4A-I; eukaryotic initiation factor 4AI; Eukaryotic initiation factor 4A-I; eukaryotic translation initiation factor 4A, isoform 1; eukaryotic translation initiation factor 4A1; initiation factor eIF-4A long form; QtsA-12280; unnamed protein product | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13681, 1973, 287436 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction