missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4A1 Recombinant Rabbit Monoclonal Antibody (23GB3835), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | eIF4A1 |
|---|---|
| Clone | 23GB3835 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088370
![]() |
Invitrogen™
MA553646 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088702
|
Invitrogen™
MA553647 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4A1 Recombinant Monoclonal antibody specifically detects eIF4A1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| eIF4A1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P60842, P60843 | |
| 13681, 1973, 287436 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3835 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ATP-dependent RNA helicase eIF4A-1; BM-010; DDX2A; Eif4; EIF4A; EIF-4A; Eif4a1; eIF-4A-I; eIF4A-I; eukaryotic initiation factor 4AI; Eukaryotic initiation factor 4A-I; eukaryotic translation initiation factor 4A, isoform 1; eukaryotic translation initiation factor 4A1; initiation factor eIF-4A long form; QtsA-12280; unnamed protein product | |
| A synthetic peptide of human eIF4A1 (1-48AA). | |
| Primary | |
| -20°C | |
| EIF4A1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title