missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4A2 Antibody (PCRP-EIF4A2-2B5), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08883AF488
This item is not returnable.
View return policy
Description
eIF4A2 Monoclonal antibody specifically detects eIF4A2 in Human samples. It is validated for ELISA, Protein Array
Specifications
| eIF4A2 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2 | |
| Recombinant full-length human eIF4A2 protein (Uniprot: Q14240) | |
| 100 μg | |
| Cancer, mTOR Pathway | |
| 1974 | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Protein Array | |
| PCRP-EIF4A2-2B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction