Learn More
EIF4A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11280-1-AP-20UL
 View more versions of this product
Product Code. 16808253
  • € 145.00 / 20µL
Description
eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.
Specifications
Specifications
EIF4A2
Polyclonal
Unconjugated
EIF4A2
BM 010, DDX2B, eIF 4A II, EIF4A, eIF4A II, EIF4A2, EIF4F
Rabbit
Antigen Affinity Chromatography
RUO
13682, 1974, 303831
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P10630, Q14240, Q5RKI1
EIF4A2
EIF4A2 Fusion Protein Ag1810
20 μL
Primary
Human, Rat, Mouse