missing translation for 'onlineSavingsMsg'
Learn More

EIF4A3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-06400F

Product Code. 18677816

  • € 407.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EIF4A3 Polyclonal antibody specifically detects EIF4A3 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EIF4A3
Polyclonal
Western Blot, Immunoprecipitation
ATP-dependent RNA helicase DDX48, ATP-dependent RNA helicase eIF4A-3, DDX48, DEAD (Asp-Glu-Ala-Asp) box polypeptide 48, DEAD box protein 48, DKFZp686O16189, EC 3.6.1, EC 3.6.4.13, eIF-4A-III, eIF4A-III, EIF4AIII, eukaryotic initiation factor 4A-III, Eukaryotic initiation factor 4A-like NUK-34, eukaryotic translation initiation factor 4A, Eukaryotic translation initiation factor 4A isoform 3, eukaryotic translation initiation factor 4A3, hNMP 265, KIAA0111eukaryotic translation initiation factor 4A, isoform 3, MGC10862, NMP 265, NMP265, Nuclear matrix protein 265, NUK34
Produced in rabbits immunized with E. coli-derived Human EIF4A3 fragment.
0.1 mL
Signal Transduction
9775
Store at 4C in the dark.
IgG
Western Blot, Immunoprecipitation
FITC
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.