missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B Antibody (1F5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37488
This item is not returnable.
View return policy
Description
eIF4B Monoclonal specifically detects eIF4B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| eIF4B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| EIF-4B, eukaryotic translation initiation factor 4B, PRO1843 | |
| Mouse | |
| 80 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 1975 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1F5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| EIF4B | |
| Purified recombinant fragment of human EIF4B expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction