Learn More
eIF4B Antibody (1F5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-37488-0.025ml
 View more versions of this product
Product Code. 18623936
  • € 203.00 / 0.03mL
Description
eIF4B Monoclonal specifically detects eIF4B in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eIF4B
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready
EIF-4B, eukaryotic translation initiation factor 4B, PRO1843
Mouse
80 kDa
0.025 mL
Core ESC Like Genes, mTOR Pathway, Stem Cell Markers
1975
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1F5
Unconjugated
PBS with 0.05% Sodium Azide
EIF4B
Purified recombinant fragment of human EIF4B expressed in E. Coli.
Affinity Purified
RUO
Primary
Human, Mouse
IgG2b