missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4B [p Ser406] Antibody (JE58-32), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32762
This item is not returnable.
View return policy
Description
eIF4B Monoclonal antibody specifically detects eIF4B in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| eIF4B | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| EIF-4B, eukaryotic translation initiation factor 4B, PRO1843 | |
| Synthetic peptide corresponding to residues surrounding Ser406 of human eIF4B protein. (Uniprot: P23588) | |
| 100 μL | |
| Core ESC Like Genes, mTOR Pathway, Phospho Specific, Stem Cell Markers | |
| 1975 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| JE58-32 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction