missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF4B Polyclonal Antibody

Product Code. 16588261
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16588261 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16588261 Supplier Invitrogen™ Supplier No. PA592449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SQSTRAASIF GGAKPVDTAA REREVEERLQ KEQEKLQRQL DEPKLERRPR ERHPSWRSEE TQERERSRTG SESSQTGTST TSSRNARRRE SEKSLENETL NKEEDCHSPT SKPPKPDQPL KVMPAPPPKE NAWVKRSSNP PARSQSSDTE QQSPTSGGGK VAPAQPSEEG PGRKDENKVD GMNAPKGQTG NSSRGPGDGG NRDHWKESDR KDGKKDQDSR SAPEPKKPEE NPASKFSSAS KYAALSVDGE DENEGEDYAE; Positive Samples: 22Rv1, HeLa, 293T, M21.

The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4B
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Eif4b
Gene Accession No. P23588, Q8BGD9
Gene Alias 2310046H11Rik; AL024095; C85189; Eif4a2; EIF4B; eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609; hypothetical protein; PRO1843
Gene Symbols Eif4b
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 352-611 of human EIF4B (NP_0014082).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1975, 75705
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.