missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98868-50ul
This item is not returnable.
View return policy
Description
eIF4E Polyclonal antibody specifically detects eIF4E in Human samples. It is validated for Western Blot
Specifications
| eIF4E | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human eIF4E. | |
| 50 μg | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway | |
| 1977 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction