missing translation for 'onlineSavingsMsg'
Learn More

eIF4E Antibody (PCRP-EIF4E-1D3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08884AF594

Product Code. 18594105

  • € 608.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

eIF4E Monoclonal antibody specifically detects eIF4E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

eIF4E
Monoclonal
Alexa Fluor 594
CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein
Recombinant full-length human eIF4E protein (Uniprot: P06730)
100 μg
Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway
1977
Store at 4°C in the dark.
IgG2a
Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
PCRP-EIF4E-1D3
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.