missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4E Recombinant Rabbit Monoclonal Antibody (HL1553)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547094
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
eIF4E is involved in several cellular processes including enhanced translational efficiency, splicing, mRNA stability, and RNA nuclear export. All eukaryotic cellular mRNAs are blocked at their 5-prime ends with the 7-methylguanosine cap structure, m7GpppX (where X is any nucleotide). EIF4E is a eukaryotic translation initiation factor involved in directing ribosomes to the cap structure of mRNAs. It is a 24-kD polypeptide that exists as both a free form and as part of a multiprotein complex termed EIF4F. The EIF4E polypeptide is the rate-limiting component of the eukaryotic translation apparatus and is involved in the mRNA-ribosome binding step of eukaryotic protein synthesis. The other subunits of EIF4F are a 50-kD polypeptide, termed EIF4A, that possesses ATPase and RNA helicase activities, and a 220-kD polypeptide, EIF4G.
Specifications
| eIF4E | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P06730, P63073, P63074 | |
| Eif4e | |
| Recombinant protein encompassing a sequence within the center region of human eIF4E. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL1553 | |
| Unconjugated | |
| Eif4e | |
| AUTS19; CBP; EG668879; EIF4E; EIF-4E; EIF4E1; EIF4EL1; Eif4e-ps; EIF4F; eIF-4F 25 kDa subunit; eukaryotic translation initiation factor 4E; eukaryotic translation initiation factor 4E-like 1; eukaryotic translation initiation factor 4E-like protein; eukaryotic translation initiation factor 4-like protein; If4e; MGC111573; mRNA cap-binding protein; OTTHUMP00000219703; OTTHUMP00000219704; OTTHUMP00000219705; translation initiation factor eIF-4E | |
| Rabbit | |
| Protein A | |
| RUO | |
| 117045, 13684, 1977 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction