Learn More
EIF4E2 Monoclonal Antibody (OTI4E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502431S
Encompass_Preferred
 View more versions of this product
Product Code. 17536881
  • € 242.00 / 30µL
Description
EIF4E2 Monoclonal antibody specifically detects EIF4E2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
EIF4E2
Monoclonal
0.56 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O60573
Mouse
Affinity chromatography
RUO
9470
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI4E7
Unconjugated
EIF4E2
2700069E09Rik; 4EHP; 4E-LP; AI036339; AV129531; D0H0S6743E; EGK_04904; eIF4E type 2; eIF-4E type 2; Eif4e2; eIF4E-2; EIF4EL3; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; eukaryotic translation initiation factor 4E family member 2; eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; IF4e; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3
Full length human recombinant protein of human EIF4E2 produced in HEK293T cell.
30 μL