missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4E2, Mouse anti-Human, Clone: 1F3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009470-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MNNKFDALKDDDSGDHDQNEENSTQKDGEKEKTERDKNQSSSKRKAVVPGPAEHPLQYNYTFWYSRRTPGRPTSSQSYEQNIKQIGTFASVEQFWRFYSHMVRPGDLTGHSDFHLFKEGIKPMWEDDANKNGGKWIIRLRKGLASRCWENLILAMLGEQFMVGEEICGAVVSVRFQEDIISIWNKTASDQATTARIRDTLRRVLNLPPNTIMEYKTHTDSIKMPGRLGPQRLLFQNLWKPRLNVPSpecifications
| EIF4E2 | |
| Monoclonal | |
| Unconjugated | |
| eukaryotic translation initiation factor 4E family member 2 | |
| 4E-LP/4EHP/EIF4EL3/IF4e | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1F3 | |
| In 1x PBS, pH 7.4 | |
| BC005874 | |
| EIF4E2 | |
| EIF4E2 (AAH05874, 1 a.a. ∼ 245 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9470 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction