missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF4E3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17834520
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17834520 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17834520 Supplier Invitrogen™ Supplier No. PA5140002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LLATIGEQFT DCAAADDEVI GVSVSVRDRE DVVQVWNVNA SLVGEATVLE KIY Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4E3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eif4e3
Gene Accession No. Q8N5X7
Gene Alias 1300018P11Rik; AI451927; eIF4E type 3; eIF-4E type 3; Eif4e3; eIF-4E3; eIF4E-3; eukaryotic translation initiation factor 4E class III; eukaryotic translation initiation factor 4E family member 3; eukaryotic translation initiation factor 4E member 3; eukaryotic translation initiation factor 4E type 3; eukaryotic translation initiation factor-3; wu:fc31f11; wu:fd15f11; zgc:92189; zgc:92189 protein
Gene Symbols Eif4e3
Host Species Rabbit
Immunogen Recombinant protein fragment of Human EIF4E3 (Product #RP-106878)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.