missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4E3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550954
This item is not returnable.
View return policy
Description
EIF4E3 Polyclonal Antibody detects endogenous levels of EIF4E3 protein.
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
Specifications
| eIF4E3 | |
| Polyclonal | |
| Unconjugated | |
| Eif4e3 | |
| 1300018P11Rik; AI451927; eIF4E type 3; eIF-4E type 3; Eif4e3; eIF-4E3; eIF4E-3; eukaryotic translation initiation factor 4E class III; eukaryotic translation initiation factor 4E family member 3; eukaryotic translation initiation factor 4E member 3; eukaryotic translation initiation factor 4E type 3; eukaryotic translation initiation factor-3; wu:fc31f11; wu:fd15f11; zgc:92189; zgc:92189 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 297481, 317649, 66892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N5X7, Q9DBB5 | |
| Eif4e3 | |
| Synthesized peptide derived from the Internal region of human eIF4E3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction