missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4E3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541955
This item is not returnable.
View return policy
Description
Peptide sequence: VQVWNVNASL VGEATVLEKI YELLPHITFK AVFYKPHEEH HAFEGGRGKH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
Specifications
| eIF4E3 | |
| Polyclonal | |
| Unconjugated | |
| Eif4e3 | |
| 1300018P11Rik; AI451927; eIF4E type 3; eIF-4E type 3; Eif4e3; eIF-4E3; eIF4E-3; eukaryotic translation initiation factor 4E class III; eukaryotic translation initiation factor 4E family member 3; eukaryotic translation initiation factor 4E member 3; eukaryotic translation initiation factor 4E type 3; eukaryotic translation initiation factor-3; wu:fc31f11; wu:fd15f11; zgc:92189; zgc:92189 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 317649 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N5X7 | |
| Eif4e3 | |
| Synthetic peptide directed towards the middle region of human EIF4E3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction