missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4ENIF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598908
This item is not returnable.
View return policy
Description
eIF4ENIF1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| eIF4ENIF1 | |
| Polyclonal | |
| Unconjugated | |
| EIF4ENIF1 | |
| 2610509L04Rik; 4E-T; A930019J01Rik; AA410001; AU021239; Clast4; D11Ertd166e; eIF4E transporter; EIF4ENIF1; eIF4E-transporter; Eukaryotic translation initiation factor 4E nuclear import factor 1; eukaryotic translation initiation factor 4E transporter; Eukaryotic translation initiation factor 4E transporter-like protein; FLJ21601; FLJ26551; OTTHUMP00000063276; RGD1560908 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 56478, 74203 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9EST3, Q9NRA8 | |
| EIF4ENIF1 | |
| Recombinant Human Eukaryotic translation initiation factor 4E transporter protein (340-630AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction