missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF4G3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA589943

Product Code. 16527621

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HKEAKIEEQE EQRKVQQLMT KEKRRPGVQR VDEGGWNTVQ GAKNSRVLDP SKFLKITKPT IDEKIQLVPK AQLGSWGKGS SGGAKASETD ALRSSASSLN RFSALQPPAP SGSTPSTPVE FDSRRTLTSR GSMGREKNDK PLPSATARPN TFMRGGSSKD LLDNQSQEEQ RREMLETVKQ LTGGVDVERN STEAERNKTR E; Positive Samples: LO2, HeLa.

The protein encoded by this gene is thought to be part of the eIF4F protein complex, which is involved in mRNA cap recognition and transport of mRNAs to the ribosome. Interestingly, a microRNA (miR-520c-3p) has been found that negatively regulates synthesis of the encoded protein, and this leads to a global decrease in protein translation and cell proliferation. Therefore, this protein is a key component of the anti-tumor activity of miR-520c-3p.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

eIF4G3
Polyclonal
Unconjugated
EIF4G3
1500002J22Rik; 4833436O05; 4930523M17Rik; eIF4G 3; eIF-4G 3; Eif4g3; eIF-4-gamma 3; eIF-4-gamma II; eIF4GII; eukaryotic translation initiation factor 4 gamma 3; eukaryotic translation initiation factor 4 gamma, 3; G1-419-52; repro8; RP11-190H11.1
Rabbit
Affinity Chromatography
RUO
8672
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.74 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
O43432
EIF4G3
Recombinant fusion protein containing a sequence corresponding to amino acids 1000-1200 of human EIF4G3 (NP_0037512).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.