missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583149
This item is not returnable.
View return policy
Description
Immunogen sequence: KDTDKFKGFC YVEFDEVDSL KEALTYDGAL LGDRSLRVDI AEGRKQDKGG FGFRKGGPDD RGMGSSRESR GGWDSRDDFN SG Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 99%.
This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| eIF4H | |
| Polyclonal | |
| Unconjugated | |
| EIF4H | |
| AU018978; D5Ertd355e; E430026L18Rik; Ef4h; EIF4H; eIF-4H; Eukaryotic translation initiation factor 4H; KIAA0038; mKIAA0038; WBSCR1; Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosomal region 1 protein homolog; Williams-Beuren syndrome chromosome region 1; Williams-Beuren syndrome chromosome region 1 homolog; WSCR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22384, 288599, 7458 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15056, Q5XI72, Q9WUK2 | |
| EIF4H | |
| Recombinant protein corresponding to Human EIF4H. Recombinant protein control fragment (Product #RP-95110). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction