missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-96987-100ul
This item is not returnable.
View return policy
Description
eIF5A2 Polyclonal antibody specifically detects eIF5A2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| eIF5A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-1 μL/mg of lysate | |
| eIF-5A-2, EIF-5A2, eIF5AII, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 2, eukaryotic translation initiation factor 5A2, eukaryotic translation initiation factor 5A-2 | |
| Produced in rabbits immunized with purified, recombinant Human eIF5A2 (Uniprot#: Q9GZV4; Met1-Lys153) | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 56648 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction