missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38116-20ul
This item is not returnable.
View return policy
Description
eIF5A2 Polyclonal antibody specifically detects eIF5A2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| eIF5A2 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| eIF-5A-2, EIF-5A2, eIF5AII, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 2, eukaryotic translation initiation factor 5A2, eukaryotic translation initiation factor 5A-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-153 of human eIF5A2 (NP_065123.1).,, Sequence:, MADEIDFTTGDAGASSTYPMQCSALRKNGFVVLKGRPCKIVEMSTSKTGKHGHAKVHLVGIDIFTGKKYEDICPSTHNMDVPNIKRNDYQLICIQDGYLSLLTETGEVREDLKLPEGELGKEIEGKYNAGEDVQVSVMCAMSEEYAVAIKPCK | |
| 20 μL | |
| Cancer, Tumor Suppressors | |
| 56648 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction