missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF5A2 Monoclonal Antibody (OTI2E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505083S
This item is not returnable.
View return policy
Description
EIF5A2 Monoclonal antibody specifically detects EIF5A2 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| EIF5A2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q93062 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11030 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI2E9 | |
| Unconjugated | |
| Rbpms | |
| 9630038B20; EIF5A2; eIF-5A2; eIF-5A-2; eIF5AII; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 2; eukaryotic translation initiation factor 5A2; Eukaryotic translation initiation factor 5A-2 | |
| Full length human recombinant protein of human RBPMS produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction