missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELA3B Mouse anti-Human, Mouse, Rat, Clone: CELA3B/1257, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14788.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against partial recombinant human ELA3B.
- High specificity
- High reactivity
Specifications
| ELA3B | |
| Monoclonal | |
| Unconjugated | |
| ELISA (2-4 ug/mL for coating) Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined | |
| P08861 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 23436 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| CELA3B/1257 | |
| Mouse monoclonal antibody raised against partial recombinant human ELA3B. | |
| In 10 mM PBS. | |
| ELA3B | |
| Recombinant protein corresponding to amino acids 82-238 of human ELA3B. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction