missing translation for 'onlineSavingsMsg'
Learn More

Elagolix sodium, MedChemExpress

Product Code. 16412187
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412187

Brand: Medchem Express HY14369

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elagolix sodium is a human GnRH receptor (GnRHR) antagonist with an IC50 and Ki of 0.25 and 3.7 nM, respectively.

Elagolix sodium is a human GnRH receptor (GnRHR) antagonist with an IC50 and Ki of 0.25 and 3.7 nM, respectively.

Specifications

Chemical Name or Material Elagolix sodium
CAS 832720-36-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C32H29F5N3NaO5
Quantity 50 mg
Synonym NBI-56418 sodium
Solubility Information H2O : ≥ 50 mg/mL (76.50 mM) ∣DMSO : 50 mg/mL (76.50 mM; Need ultrasonic)
SMILES O=C(O[Na])CCCN[C@H](C1=CC=CC=C1)CN(C(N(CC2=C(C(F)(F)F)C=CC=C2F)C(C)=C3C4=CC=CC(OC)=C4F)=O)C3=O
Molecular Weight (g/mol) 653.57
Formula Weight 653.57
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.