missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elastin Antibody (ELN/3131R), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08890UV
This item is not returnable.
View return policy
Description
Elastin Monoclonal antibody specifically detects Elastin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Elastin | |
| Monoclonal | |
| DyLight 350 | |
| elastin, FLJ38671, FLJ43523, SVAS, tropoelastin, WBS, WS | |
| Recombinant full-length human Elastin protein (Uniprot: P15502) | |
| 100 μg | |
| Extracellular Matrix | |
| 2006 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| ELN/3131R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction