missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Elastin Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody
€ 185.00 - € 415.00
Specifications
| Antigen | Elastin |
|---|---|
| Clone | C4 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17938732
|
Invitrogen™
MA551168 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17920853
![]() |
Invitrogen™
MA551167 |
200 μL |
€ 415.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| Elastin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 5830419M17Rik; AI385707; AI480567; AW229038; BB105748; E030024M20Rik; elastin; elastin (supravalvular aortic stenosis, Williams-Beuren syndrome); elastin a precursor; elastin b precursor; elastin c precursor; elastin microfibril interface located protein 1; elastin microfibril interface-located protein 1; Elastin microfibril interfacer 1; Eln; EMI; EMILIN; EMILIN1; EMILIN-1; FLJ38671; FLJ43523; gp115; RATTREL11; SVAS; TREL11; Trela; Trela26; Tropoelastin; WBS; WS | |
| ELN | |
| IgG2b κ | |
| Protein A/G | |
| ELN |
| C4 | |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P15502, Q99372 | |
| 2006, 25043 | |
| Recombinant Human ELN protein, Gly392-Ala645 (Accession #P15502). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title