Learn More
Hanna Instruments™ Electrolyte Fill Solution
Click to view available options
Junction:
Double
Single
Concentration:
3.5M KCl
3.5M KCl + AgCl
Unit Size:
120mL
Each
Description
pH and ORP electrode fill solution that contains 3.5M KCl which is specifically made for double junction electrodes and 3.5M KCl + AgCl specifically for single junction electrodes
- Convenient 30 mL bottles
- Clearly marked bottle with expiration date and lot number
- Four bottles of 30 mL 3.5M KCl (+ AgCl for single junction) electrolyte solution that ensures optimum measurement performance from your single junction pH or ORP electrode.
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Concentration | 3.5M KCl |
| Type | Electrolyte Fill Solution |
| Junction | Double |
| Quantity | 4 Pack |