Learn More
Hanna Instruments™ Electrolyte Fill Solution
Product Code. p-6895007
Click to view available options
Junction:
Double
Single
Concentration:
3.5M KCl
3.5M KCl + AgCl
Unit Size:
120mL
Each
Product Code. 10468442
Brand: Hanna Instruments™ HI7071
Only null left
Add to basket
Add to basket
Contains 3.5M KCl for double junction electrodes and 3.5M + AgCl for single junction. Hanna Instruments™ Electrolyte Fill Solution ensures optimum measurement performance from a single junction pH or ORP electrode.
pH and ORP electrode fill solution that contains 3.5M KCl which is specifically made for double junction electrodes and 3.5M KCl + AgCl specifically for single junction electrodes
  • Convenient 30 mL bottles
  • Clearly marked bottle with expiration date and lot number
  • Four bottles of 30 mL 3.5M KCl (+ AgCl for single junction) electrolyte solution that ensures optimum measurement performance from your single junction pH or ORP electrode.
Specifications
Capacity (Metric) 30 mL
Concentration 3.5M KCl + AgCl
Type Electrolyte Fill Solution
Filling Solution Type Electrolyte
Junction Single
Quantity 4 Pack
compliance-icons
Product Identifier
  • 3.5M KCl with AgCl Reference Electrolyte
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : silver chloride